PEEK Tubing
PEEK Tubing
Actual product may vary
PEEK Tubing
Useful for when contact between the sample and metal components must be avoided in an HPLC system, such as when working with metal complexing agents or certain biochemical compounds
Have Questions?
Catalog NumberDiameter (English) InnerDiameter (English) Outer
549940.055 in.1/16 in.
Catalog number 54994
Price (MYR)
-
Diameter (English) Inner:
0.055 in.
Diameter (English) Outer:
1/16 in.
  • A popular replacement for stainless steel tubing in various places in the HPLC system
  • Useful when contact between the sample and metal components must be avoided, such as when working with metal complexing agents or certain biochemical compounds
  • Tubing is flexible, easy to cut, has excellent mechanical stability and chemical compatibility
  • Tetrahydrofuran (THF), dimethyl sulfoxide (DMSO), methylene chloride, and concentrated nitric and sulfuric acids should not be used with PEEK tubing
  • 1/16 in. tubing is color coded for easy identification of the internal diameter
  • Maximum Temperature: 100°C
Specifications
TypeTubing
ColorNatural
For Use With (Application)HPLC Systems
Diameter (English) Inner0.055 in.
Diameter (English) Outer1/16 in.
Overall Length (English)10 ft.
Overall Length (Metric)3 m
MaterialPEEK
Maximum Pressure1000 psi
Max. Operating Temperature (Metric)100°C
Unit SizeEach