Titan3™ Regenerated Cellulose Syringe Filters
Titan3™ Regenerated Cellulose Syringe Filters
Actual product may vary
Titan3™ Regenerated Cellulose Syringe Filters
Titan3™ Regenerated Cellulose Syringe Filters
Titan3™ Regenerated Cellulose Syringe Filters
Titan3™ Regenerated Cellulose Syringe Filters
With a robust design that includes a reinforcing outer ring configuration and integral pre-filter, Titan3 Syringe Filters are an ideal choice for high solids applications or where maximum confidence is required.
Have Questions?
Change viewbuttonViewtableView
Catalog Number
54513-RC
52204-RC
also known as NSC GROUP 52204-RC
52213-RC
54525-RC
Catalog number 54513-RC
View Price:Sign in to see your account pricing. Need an account? Register with us today.
With a robust design that includes a reinforcing outer ring configuration and integral pre-filter, Titan3 Syringe Filters are an ideal choice for high solids applications or where maximum confidence is required.
Outstanding Performance
  • Low extractable membranes and housings ensure that no filter-borne contaminants are added to your sample
  • Performance-tested under actual HPLC conditions
  • Integral 1mm borosilicate glass pre-filter (30mm devices) ensures greater confidence with high solids samples
  • Reinforcing ring provides greater strength to the housing to prevent leakage and bursting (30mm products pressure rated to 120 psi)
  • Housings are manufactured from solvent-resistant, low-extractable polypropylene resins specifically selected for wide compatibility with common HPLC sample matrices
  • Able to filter solutions at temperatures up to 100°C
Convenient and Easy to Use
  • Color coding makes it easy to select the correct membrane and pore size
  • Packed in re-usable rigid transparent color-coded containers
  • Syringe filters can be sterilized by autoclave at 125°C for 15 minutes
  • Enhanced Luer lock inlet prevents leakage; outlet fitting is a standard size male Luer-slip fitting for ease of filtrate collection
Specifications
Certifications/ComplianceTitan3 Polypropylene Syringe Filter Housings Meet The Requirements of 21 CFR 177.1520. Membrane: HPLC Certified Regenerated Cellulose.
ColorLight Brown
DisposableSingle-use
Material (Membrane)Regenerated Cellulose
Pore Size0.45 μm
SterilityNon-sterile
For Use With (Application)Sample Filtration: low nonspecific binding, tissue culture media, general biological samples
Hold up Volume (After Air Purge)<35 μL
Housing MaterialPolypropylene
Membrane TypeRegenerated Cellulose
PrefilterNo
Pressure Rating115 psi
Product LineTitan3
Quantity200/Pk.
TypeSyringe Filter
Volume (Metric)<10 mL
Diameter (Metric)17 mm
Diameter (Metric) Membrane17 mm
Unit SizeCase