Specifications
DescriptionTFT-80.2 Fixed Angle Rotor
MaterialTitanium
TypeFixed Angle Rotor
Capacity12 x 2 mL
Max. RCF429,643 x g
Max. Speed80,000 rpm
Radius (Metric)60.1 mm
Radius (Metric) Min.33.9 mm
K Factor22.6
Max. Tube Length51 mm
Max. Tube Diameter8 mm
Net Weight5.5 lb. (2.5 kg)
For Use With (Equipment)Thermo Scientific™ Sorvall™ WX ultraCentrifuges
Angle25°
Avg. RCF335,994 x g
Min. RCF242,344 x g
Unit SizeEA