AH-629 (36mL) Swinging Bucket Rotor
AH-629 (36mL) Swinging Bucket Rotor
Actual product may vary
AH-629 (36mL) Swinging Bucket Rotor
Facilitate large volume particle isolations using Thermo Scientific™ AH-629 6 x 36mL Swinging-Bucket Rotor.
Have Questions?
Catalog NumberCapacity
542846 x 36 mL
Catalog number 54284
Price (MYR)
109,888.00
EA
Add to cart
Capacity:
6 x 36 mL
Price (MYR)
109,888.00
EA
Add to cart
Facilitate large volume particle isolations using Thermo Scientific™ AH-629 6 x 36mL Swinging Bucket Rotor.
Specifications:
  • Swinging bucket aluminum made rotor
  • Maximum capacity: 6 × 36mL (0.21L)
  • Max. speed/RCF: 29,000rpm/151,243 x g
  • K factor: 242
  • Angle: 90°
Ordering Information:
Order 17mL buckets (six) (52030), ultracrimp/clearcrimp sealing tool (03920), crimp gauge replacement (03919), extra plugs and caps (03047), tube removal tool (12826), tube racks (two) (03539) separately. Contact your sales representative for information.
Compatible with:
Thermo Scientific™ Sorvall™ WX Ultra Series Floor Model Centrifuge
Recommended for:
Large volume viral and subcellular particle isolations, RNA pelleting.
Specifications
DescriptionAH-629 (36mL) Swinging Bucket Rotor
MaterialAluminum
TypeSwinging Bucket Rotor
Capacity6 x 36 mL
Max. RCF151,243 x g
Max. Speed29,000 rpm
Radius (Metric)161 mm
Radius (Metric) Min.72.1 mm
K Factor242
Max. Tube Length89 mm
Max. Tube Diameter25 mm
Net Weight14.1 lb. (6.4 kg)
For Use With (Equipment)Thermo Scientific™ Sorvall™ WX ultraCentrifuges
Angle90°
Avg. RCF109,487 x g
Min. RCF67,731 x g
Unit SizeEA