Nalgene™ 230mm Ceramic Metal Desiccator Plate
Nalgene™ 230mm Ceramic Metal Desiccator Plate
Actual product may vary
Nalgene™ 230mm Ceramic Metal Desiccator Plate
Ensure greater resistance to thermal shock than porcelain using this guaranteed unbreakable and corrosion-resistant ceramic metal desiccator plate.
Have Questions?
Catalog NumberDiameter (Metric) Outer
5312-0230230 mm
Catalog number 5312-0230
Price (MYR)
3,140.00
EA
Add to cart
Diameter (Metric) Outer:
230 mm
Price (MYR)
3,140.00
EA
Add to cart
Ensure greater resistance to thermal shock than porcelain using the guaranteed unbreakable and corrosion-resistant Thermo Scientific™ Nalgene™ 230mm Ceramic Metal Desiccator Plate.
  • 230mm diameter fits the Nalgene Classic Dessicator (5311-0250) as well as the 250mm Nalgene Blue Desiccators (5309-0250 and 5310-0250)
  • Numbered quadrants for easy location of crucibles and other containers
  • Six holes in each quadrant and a 3mm center hole to enhance airflow and drying efficiency
  • Autoclavable
Ordering Information:
Individually packaged
Please recycle the corrugated cartons and LDPE bags in which your Nalgene products are packaged.
Specifications
Diameter (Metric) Outer230 mm
No. per Pack1
No. per Case6
DescriptionDesiccator Plate Ceramic/Metal 230mm
Diameter (English) Outer9.1 in.
Unit SizeEA