Wellwash™ Versa Microplate Washer
Wellwash™ Versa Microplate Washer
Actual product may vary
Wellwash™ Versa Microplate Washer
Wash 96- or 384-well plates securely with the Thermo Scientific™ Versa™ Microplate Washer, an easy-to-use microplate-strip washer for routine ELISA applications.
Have Questions?
Catalog NumberModel
5165050Wellwash Versa 2x12
Catalog number 5165050
Price (MYR)
55,342.00
EA
Add to cart
Model:
Wellwash Versa 2x12
Price (MYR)
55,342.00
EA
Add to cart
Wash 96- or 384-well plates securely with the Thermo Scientific™ Wellwash™ Versa Microplate Washer, an easy-to-use microplate-strip washer for routine ELISA applications. Designed for high performance and versatility, enjoy the ease of use and convenience provided through a graphical interface, local language version and USB port.
Simplify your ELISA workflow by using our eGuide to see our complete portfolio for all your ELISA needs.
Easy and convenient to use
  • Large color display and keypad for easy, convenient use
  • Clear display shows everything on one screen
  • User interface is logic and intuitive, allowing quick protocol development
  • Easy-to-use software requires minimal training for fast startup
Reliable and secure performance
  • Unpressurized bottles are safe and secure to use
  • Liquid level sensors are present in both wash and waste bottles
  • Automatic rinse and prime features assure safe performance
  • Aerosol cover prevents aerosolization of infectous diseases
  • Plate sensor recognizeds if plate is present
Optimal washing performance
  • Sweep mode guarantees an extremely low residual volume in the well, resulting in reliable assay results
  • Washing performance is further optimized by adjustable parameters such as dispense and aspiration height, and aspiration speed
Added versatility
  • A USB port for transferring protocols between several washers and for printing reports and log files
  • Specially designed 2x8 cell wash head is available for cell washing
  • Two 1x8 wash heads, two wash bottles and one rinse bottle, optional two 1x12 wash heads, 1x16 wash heads and 2x8 cell wash head offer increased flexibility
  • Possibility to connect to automation systems
Includes:
2x2L wash bottles, 1x2L rinse bottle, 1x4L waste bottle, two 1x8 wash heads, aerosol cover
Recommended for:
ELISA-based assays, cell washing
Warranty: 1 year
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Accuracy< 5% @ 300μL, with n x 8 or n x 12 wash head; < 5% @ 250μL, with 2 x 8 cell wash head; < 5% @ 100μL, with 1 x 16 wash head
Certifications/ComplianceCE Marked
ConnectionsUSB
Wash/Waste Bottle Capacity3
BottlesWash Bottle (2 x 2 L), Rinse Bottle (1 x 2 L), Waste Bottle (1 x 4 L)
Precision< 3% (CV) @ 300μL, with n x 8 or n x 12 wash heads
Dispense Volume50 to 400 μL (96); 20 to 120 μL (384)
Wash HeadsUp to two 1 x 12 wash heads and optional up to two 1 x 8 wash heads and 2 x 8 cell wash head for 96 well plate; 1 x 16 wash head for 384 well plate
Plate Type96-well Plate, 384-well Plate
Residual Volume<2 μL
Weight (Metric)9 kg
Wash Volume50 to 1000 μL (96); 20 to 300 μL (384)
Rinse Volume5 to 100 mL
DisplayLCD
For Use With (Application)ELISA-Based Assays; Cell Washing
Frequency50/60 Hz
ModelWellwash Versa 2x12
Cell WashingYes
Includes2x2L wash bottles, 1x2L rinse bottle, 1x4L waste bottle, two 2x12 wash heads, and aerosol cover
Width (English)15.2 in.
Width (Metric)385 mm
Height (English)13.6 in.
Height (Metric)345 mm
Depth (English)9.4 in.
Depth (Metric)240 mm
Automation InterfaceYes (USB or RS-232C)
Weight (English)20 lb.
Electrical Requirements100/240 V 50/60 Hz
Voltage100/240 V
Prime Volume5 to 100 mL
Unit SizeEA
Frequently asked questions (FAQs)
What accessories are available for use with my washer? Can I order extra bottles?
Yes. Please refer to the product brochure on the following page for a complete list of available accessories: https://www.thermofisher.com/us/en/home/life-science/lab-equipment/microplate-instruments/microplate-washers.html
Is my washer integratable with robotics?
Wellwash Versa is integrateable; basic Wellwash is not.
What are the dimensions / weight of the washers?
34.5 cm x 38.5 cm x 24.0 cm; 9 kg
What dispense volumes are allowed?
50-400 μL for 96 well plates, 20-120 μL for 384 well plates (384-well on Versa only) for dispense volumes are allowed.
What wash volumes are allowed?
50-1000 μL for 96 well plates, and 20-300 μL for 384 well plates (384-well on Versa only) wash volumes are allowed.