Wellwash™ Microplate Washer
Wellwash™ Microplate Washer
Actual product may vary
Wellwash™ Microplate Washer
Wash 96-well plates securely with this easy-to-use, reliable microplate-strip washer for routine ELISA applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberModel
5165040Wellwash 1x12
5165000Wellwash 1x8
Catalog number 5165040
Price (MYR)
39,488.00
EA
Add to cart
Model:
Wellwash 1x12
Price (MYR)
39,488.00
EA
Add to cart
Wash 96-well plates securely with the Thermo Scientific™ Wellwash™ Microplate Washer, an easy-to-use, reliable microplate-strip washer for routine ELISA applications. The Wellwash features a graphical user interface, local language versions and USB port for optimal ease-of-use and convenience.
Simplify your ELISA workflow by using our eGuide to see our complete portfolio for all your ELISA needs.
Features:
  • Large color clear display and keypad for easy, convenient use
  • User interface is logic and intuitive for quick protocols development
  • Easy-to-use software requires minimal training for fast startup
  • Unpressurized bottles are safe and secure to use
  • Liquid level sensors are present in both wash and waste bottle for security
  • Automatic prime feature assure safe performance
  • Aerosol cover prevents aerosolization of infectous diseases
  • Plate sensor recognizes if plate is present
  • Wellwash is designed to efficiently remove the excess liquid from the wells
  • Low residual volumes provide optimal washing performance and reliable assay results
Includes:
1x2L wash bottle, 1x2L waste bottle, 1x8L wash head, aerosol cover
Recommended for:
ELISA-based assays
Warranty: 1 year
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Accuracy< 5% @ 300μL
Certifications/ComplianceCE Marked
ConnectionsUSB
Wash/Waste Bottle Capacity1
BottlesWash Bottle (2 L), Waste Bottle (2 L)
Precision< 3% (CV) @ 300μL (96-well plate)
Dispense Volume50 to 400 μL
Wash Heads1 x 12
Plate Type96-Well Plates
Residual Volume<2 μL
Weight (Metric)8 kg
Wash Volume50 to 1000 μL
DisplayLCD
For Use With (Application)ELISA-Based Assays
Frequency50/60 Hz
ModelWellwash 1x12
Cell WashingNo
Includes1 x 12 wash head, 1 x 2L wash bottle, 1 x 2L waste bottle, aerosol cover
Width (English)15.2 in.
Width (Metric)385 mm
Height (English)13.6 in.
Height (Metric)345 cm
Depth (English)9.4 in.
Depth (Metric)240 mm
Automation InterfaceNo
Weight (English)17.6 lb.
Electrical Requirements100/240 V 50/60 Hz
Voltage100/240 V
Prime Volume5 to 100 mL
Unit SizeEA
Frequently asked questions (FAQs)
What accessories are available for use with my washer? Can I order extra bottles?
Yes. Please refer to the product brochure on the following page for a complete list of available accessories: https://www.thermofisher.com/us/en/home/life-science/lab-equipment/microplate-instruments/microplate-washers.html
Is my washer integratable with robotics?
Wellwash Versa is integrateable; basic Wellwash is not.
What are the dimensions / weight of the washers?
34.5 cm x 38.5 cm x 24.0 cm; 9 kg
What dispense volumes are allowed?
50-400 μL for 96 well plates, 20-120 μL for 384 well plates (384-well on Versa only) for dispense volumes are allowed.
What wash volumes are allowed?
50-1000 μL for 96 well plates, and 20-300 μL for 384 well plates (384-well on Versa only) wash volumes are allowed.