Search Thermo Fisher Scientific
Catalog Number | Chamber Material | Voltage | CO2 Sensor Technology |
---|---|---|---|
51033589 | Electropolished Stainless Steel | 230 V | TC Sensor |
Achieve your next breakthrough. The Thermo Scientific™ Heracell™ VIOS CO2 incubator is specifically designed to deliver the performance reliability, ease of operation, and value required to support a range of culturing needs from basic research to the demanding, leading-edge applications, so you're ready for whatever comes next.
Better solutions for optimal cell growth. Our CO2 incubators with non-corrosive stainless-steel interior chambers provide the ideal in-vitro environment. Large 255L capacity provides space for high throughput applications and accommodates spaces for culture devices for larger sample output.