Heratherm™ Refrigerated Incubators
Heratherm™ Refrigerated Incubators
Actual product may vary
Heratherm™ Refrigerated Incubators
Offers precise temperature control, increased energy efficiency, and ease of usability. Equipped with Peltier technology to reach and maintain the precise ambient temperatures you need for reliable results.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Voltage
51031567381 L230 V
51031564178 L230 V
51031562178 L100/230 V
Catalog number 51031567
Price (MYR)
100,819.00
EA
Add to cart
Capacity (Metric):
381 L
Voltage:
230 V
Price (MYR)
100,819.00
EA
Add to cart
Achieve untapped potential in incubation with the temperature precision you need for applications that demand it. With a temperature range of +5 to +70°C, Thermo Scientific™ Heratherm™ Refrigerated Incubators are equipped with Peltier technology to reach and maintain the precise ambient temperatures you need for reliable results. With no hazardous refrigerants, these units use less energy than the conventional product, making it the ideal choice for energy-conscious labs.
  • Temperature range +5 to +70°C
  • Choose benchtop model 178L / 6.3 cu.ft. or floor model with casters 381L / 13.5 cu. ft.
  • Great temperature uniformity and stability to keep the samples in a safe environment –> uniformity as low as +/-0.3°C (20°C), stability as low as +/-0.1°C
  • Energy consumption as low as 80Wh/h (benchtop) / 150Wh/h (floor) at 20°C - Note that comparable compressor cooled units consume up to 6 times this amount*
  • CFC free / HFC free polyurethane insulation foam for outstanding temperature performance and condensation prevention
  • Heated door with intelligent energy control to ensure no condensation – even at low temperatures
  • Sophisticated timer function with weekly, daily or real time mode
  • Intuitive programmability function for defined temperature ramping, soak/cool and dwell: store up to 10 programs with 10 easy steps
  • Adjustable over temperature shut-off for protocols that require setting of over temperature for protection – (class 2 according to norm DIN12880)
  • Safe viewing of samples through internal glass door with limited impact on temperature
  • Safe containment with automatic over and under temperature alarm
  • Standard access port for data monitoring (large diameter of 42mm / 1.65 in.)
  • Flexible shelf system for optimal loading, easy to remove for cleaning, with non-tip protection
  • Stainless steel interior (1.4301 / ASTM 304) and rounded corners for easy cleaning
*Based on comparison with models BK6160/Precision815
Note: stated values are based on norm DIN12880 testing.
Specifications
Capacity (English)13.5 cu. ft.
Certifications/ComplianceCE
DescriptionHeratherm refrigerated incubator IMP400, w/ electrical outlet, 230V, 50/60Hz
Capacity (Metric)381 L
Chamber MaterialStainless Steel (304/1.4301)
Dimensions (D x W) Shelf20.79 x 21.06 in. (528 x 498 mm)
MaterialCoated Steel
No. of Shelves2/17
Plug TypeEU plug CEE7/7
Temperature Range (Metric)5°C to 70°C
TypeAutomated Incubator
DisplayLarge Vacuum Fluorescent
Door StyleSolid outer door & internal glass door
Green FeaturesEnergy efficient
IncludesInternal Electrical Outlet: EU (CEE 7/4 plug and interior outlet)
InsulationCFC free/HFC free polyurethane insulation foam
ModelIMP400
No. of Doors1
Port Size1.6 in. (42mm)
RefrigerantPeltier technology: no refrigerant
Temperature Accuracy0.1°C
Temperature Uniformity≤0.3°C at 20°C; ≤0.7°C at 37°C
Dimensions (D x W x H) Interior524 x 544 x 1335 mm (20.6 x 21.4 x 52.6 in.)
Dimensions (L x W x H)770 x 778 x 1545 mm ( 30.3 x 30.6 x 60.8 in.)
Height (English)60.8 in. (casters not included: 7.1 in.)
Height (Metric)1545 mm (casters not included: 180 mm)
Shipping Weight (English)476 lb.
Shipping Weight (Metric)216 kg
Weight (English)368 lb.
Weight (Metric)167 kg
Electrical Requirements230 V, 50/60 Hz, 6.0 A, 1360 w