Heratherm™ General Protocol Ovens
Heratherm™ General Protocol Ovens
Actual product may vary
Heratherm™ General Protocol Ovens
Perform routine, day-to-day work in these general protocol ovens, which provide a safe, ideal heating and drying solution for research, clinical or industrial needs.
Have Questions?
Catalog NumberCapacity (Metric)TypeElectrical RequirementsDimensions (L x W x H)
51028114176 LGravity Convection208/240 V 60 Hz29.1 x 25.2 x 36.2 in. (73.8 x 64 x 92 cm)
Catalog number 51028114
Price (MYR)
20,001.00
EA
Add to cart
Capacity (Metric):
176 L
Type:
Gravity Convection
Electrical Requirements:
208/240 V 60 Hz
Dimensions (L x W x H):
29.1 x 25.2 x 36.2 in. (73.8 x 64 x 92 cm)
Price (MYR)
20,001.00
EA
Add to cart
Perform routine applications simply and safely with Thermo Scientific™ Heratherm™ General Protocol Ovens. These ovens provide an ideal heating and drying solution for research, clinical or industrial needs.
Features:
  • Operating temperatures from 50°C to 250°C
  • Automatic over-temperature alarm system
  • Low overall energy consumption
  • Built-in timer
  • Corrosion-resistant stainless steel chambers with rounded corners
  • Large, easy-to-read vacuum fluorescent display
  • Easy-to-use, microprocessor-controlled touch button operation
  • Doors can be opened at over 180°C
Ordering Information:
Availability may vary by country
Warranty: 2-year warranty
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Controller TypeMicroprocessor control with vacuum fluorescent display
Capacity (Metric)176 L
Temperature (Metric)50°C to 250°C
TypeGravity Convection
No. of Shelves2 supplied/19 max.
Temperature Stability±0.4°C
TimerON / OFF
Dimensions (D x W x H) Interior23.2 x 17.2 x 26.8 in. (58.9 x 43.8 x 68 cm)
Dimensions (L x W x H)29.1 x 25.2 x 36.2 in. (73.8 x 64 x 92 cm)
Length (English)29.1 in.
Length (Metric)73.8 cm
Width (English)25.2 in.
Width (English) Interior17.2 in.
Width (Metric)64 cm
Width (Metric) Interior43.8 cm
Height (English)36.2 in.
Height (English) Interior26.8 in.
Height (Metric)92 cm
Height (Metric) Interior68 cm
Depth (English) Interior23.2 in.
Depth (Metric) Interior58.9 cm
Electrical Requirements208/240 V 60 Hz
Voltage208/240 V
Wattage3360 w
Amperage14 A
Frequency60 Hz
Unit SizeEA