Barnstead™ Pretreatment for Point of Use Water Purification Systems
Barnstead™ Pretreatment for Point of Use Water Purification Systems
Actual product may vary
Barnstead™ Pretreatment for Point of Use Water Purification Systems
Barnstead™ Pretreatment for Point of Use Water Purification Systems
Barnstead™ Pretreatment for Point of Use Water Purification Systems
Barnstead™ Pretreatment for Point of Use Water Purification Systems
Barnstead Pretreatment for Point of Use Water Purification Systems
Have Questions?
Catalog number 50157886
Price (MYR)
9,276.00
EA
Add to cart
Price (MYR)
9,276.00
EA
Add to cart
Barnstead Pretreatment contains PreFilter, Hardness Stabilizer and Carbon Filter. The unit contains pressure regulator and pressure gauges. Space saving design comes with Wall Mount capability.
  • Use pretreatment when feed water quality doesn't match the requirements for your system
  • Contains pre-filter, hardness stabilizer and carbon filter
Specifications
Cartridge TypePretreatment Cartridge
DescriptionPretreatment includes Particle Filter, Carbon Filter and Hardness Stabilizer Catridges ( D502113, D502114 and D502115 )
TypeWater Purification System
For Use With (Equipment)Smart2Pure Pro
IncludesPressure Regulator, Pressure Gauges, Three Cartridges and Wall Mount
Dimensions (L x W x H)14.5 in x 13.3 in x 20.0 in.
Weight (English)24.5 lb.
Weight (Metric)11.4 kg
Unit SizeEA