Reverse Osmosis Cartridge
Product Image
Reverse Osmosis Cartridge
Choose the required Thermo Scientific™ Reverse Osmosis (RO) Cartridge for your Thermo Scientific™ Barnstead™ point of use water purification system, as indicated.
Have Questions?
Catalog number 50157608
Price (MYR)
2,942.00
EA
Add to cart
Price (MYR)
2,942.00
EA
Add to cart
Choose the required Thermo Scientific™ Reverse Osmosis (RO) Cartridge for your Thermo Scientific™ Barnstead™ point of use water purification system, as indicated.
Reverse Osmosis Cartridges for following Barnstead Water Purification Systems:
  • Smart2Pure™ Pro
Specifications
DescriptionReverse Osmosis Cartridge, 16L/hr
For Use With (Equipment)Smart2Pure™ Pro
TypeReverse Osmosis Cartridge
Unit SizeEA