Cell Locker™ Chamber with Copper Tray and Transport Cover
Product Image
Cell Locker™ Chamber with Copper Tray and Transport Cover
Protect your most sensitive cells, even during transport.
Have Questions?
Catalog NumberNo. of Places
50154739X66
Catalog number 50154739X6
Price (MYR)
0.00
EA
Add to cart
No. of Places:
6
Price (MYR)
0.00
EA
Add to cart
Protect your most sensitive cells, even during transport.
Each individual Cell Locker™ Chamber is effectively a quarantine chamber. Can be manually disinfected or can be autoclaved.
  • Dual 0.2 μm membrane filters permit air circulation but exclude microbial contaminants
  • Replacable 0.2 μm membrane filter is hydrophobic, oleophobic, resistant to organic solvents and tested for biosafety and low cytotoxicity
  • Dishes and flasks can be removed individually on a work tray or inside the entire chamber with optional transport cover, thus protecting samples from outside air
  • Non-skid tray slides out to allow access to culture vessels, or the entire tray can be removed
Specifications
No. of Places6
For Use With (Application)Cell Protection and Transport
IncludesChamber, Copper Tray, and Transport Cover
Quantity1 Ea.
Unit SizeEA