Cell Locker™ Chamber with Stainless Steel Tray, Stainless Steel
Cell Locker™ Chamber with Stainless Steel Tray, Stainless Steel
Actual product may vary
Cell Locker™ Chamber with Stainless Steel Tray, Stainless Steel
Protected chambers for your most sensitive cells
Have Questions?
Catalog NumberNo. of Places
5015165053
Catalog number 50151650
Price (MYR)
-
No. of Places:
53
Protected chambers for your most sensitive cells
  • Each individual Cell Locker Chamber is effectively a quarantine chamber.
  • Dual 0.2μm membrane filters permits air circulation but exclude microbial contaminants.
  • The replacable 0.2μm membrane filter is hydrophobic, oleophobic, resistant to organic solvents and tested for biosafety and low cytotoxicity.
  • Independent 3rd party tests validate protection against circulating microorganisms (when used as directed)
Each Cell Locker Chamber holds 9 each T-75 cell culture flasks, 20 each 6-well plates, or 24 each 96-well plates.
  • Dishes and flasks can be removed individually on a work tray or inside the entire Cell Locker Chamber with optional transport cover, thus protecting samples from outside air.
  • The non skid tray slides out to allow access to culture vessels, or the entire tray can be removed.
  • The Cell Locker Chamber can be manually disinfected or can be autoclaved.
Warranty: 1 year
Specifications
DescriptionCell Locker Chamber with stainless steel tray
Chamber MaterialStainless Steel
Holds9 each T-75 cell culture flasks, 20 each 6-well plates, or 24 each 96-well plates
LightingLED
MaterialStainless Steel
No. of Places53
No. of Shelves1
TypeCell Locker Chamber
Warranty1 Year
Unit SizeEA