Hand Dispenser Kit
Hand Dispenser Kit
Actual product may vary
Hand Dispenser Kit
Hand Dispenser Kit
Hand Dispenser Kit
Hand Dispenser Kit
Increase lab flexibility with the Thermo Scientific™ Hand Dispenser Kit, compatible with many Thermo Scientific™ Barnstead™ water purification systems.
Have Questions?
Catalog NumberDescription
50138221Hand Dispenser Kit
Catalog number 50138221
Price (MYR)
5,462.00
EA
Add to cart
Description:
Hand Dispenser Kit
Price (MYR)
5,462.00
EA
Add to cart
Increase flexibility in the lab with the Thermo Scientific™ Hand Dispenser Kit, compatible with many Thermo Scientific™ Barnstead™ water purification systems. This kit includes a dispenser with 3 meter cord that connects to the water system tank and a 0.2 micron final filter. It is compatible with Barnstead LabTower™, Pacific™ and Smart2Pure™ systems.
Optional for the following Barnstead water purification systems:
  • LabTower™ EDI
  • LabTower™ RO
  • LabTower™ TII
  • Smart2Pure™
  • Pacific™ TII + tank (tank must have optional pump)
  • Pacific™ RO + tank (tank must have optional pump)
Specifications
DescriptionHand Dispenser Kit
For Use With (Equipment)LabTower EDI, LabTower TII, Smart2Pure, Pacific TII + Tank (Tank Must Have Optional Pump), Pacific RO + Tank (Tank Must Have Optional Pump), LabTower RO
IncludesHand Dispenser Kit (includes hand dispenser with 3 meter cord, and 0.2 micron final filter)
Unit SizeEA