Heratherm Stacking Kit
Heratherm Stacking Kit
Actual product may vary
Heratherm Stacking Kit
Stacking kit for 2 100L or 60L on top of 100L Heratherm ovens and incubators
Have Questions?
Catalog number 50126666
Price (MYR)
2,764.00
EA
Add to cart
Price (MYR)
2,764.00
EA
Add to cart
Stacking kit for 2 100L or 60L on top of 100L Heratherm ovens and incubators. Recommended if performing decontaminiantion cycle in lower unit or stacking an incubator on an oven
Heratherm Stacking kit
For 2 100L or 60L on top of 100L Heratherm ovens and incubators
Recommended if performing decontaminiantion cycle in lower unit or stacking an incubator on an oven
Suitable for use with:
General Protocol
Advanced Protocol
Advanced Protocol Security ovens
448 x 511 mm
Specifications
Description100 L or 60 L on top of 100 L Heratherm ovens and incubators
Capacity (Metric)100 L or 60 L
TypeOven Accessories
Unit SizeEA