Additional Activated Carbon Filter
Additional Activated Carbon Filter
Actual product may vary
Additional Activated Carbon Filter
Additional Activated Carbon Filter
Additional Activated Carbon Filter
Additional exhaust carbon filters can be fitted on top of the BSC, but should only be used for traces of solvents being used as an adjunct to microbiological study.
Have Questions?
Catalog NumberFor Use With (Equipment)
50078783Retrofit to 0.9 m Herasafe KS/Safe 2020 Cabinets
Catalog number 50078783
Price (MYR)
-
For Use With (Equipment):
Retrofit to 0.9 m Herasafe KS/Safe 2020 Cabinets
Additional exhaust carbon filters can be fitted on top of the BSC, but should only be used for traces of solvents being used as an adjunct to microbiological study.
  • Additional Activated Carbon Filter
  • Fittings Included.
  • Additional exhaust carbon filters can be fitted on top of the BSC, but should only be used for traces of solvents being used as an adjunct to microbiological study.
Specifications
Certifications/ComplianceEN 12469
Product TypeFilter
Filter TypeCarbon
IncludesFittings
For Use With (Equipment)Retrofit to 0.9 m Herasafe KS/Safe 2020 Cabinets
Unit SizeEA