Heraguard™ ECO Clean Bench Accessories
Heraguard™ ECO Clean Bench Accessories
Actual product may vary
Heraguard™ ECO Clean Bench Accessories
Integrate Thermo Scientific™ Heraguard™ ECO Clean Bench Accessories with the ECO clean bench to increase the flexibility and enhance operations.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeDescriptionFor Use With (Equipment)
50052462Wind Screen Protection KitWindscreen protection kitHeraguard ECO 1.8
50044679Media ValvesWater tap G3/8 connection (Black) with long neck for installation in side wallsHeraguard ECO 0.9, 1.2, 1.5, 1.8, 1.8/95
51901073UV OptionsUV Option, factory installed, comes with UV-resistant Night CoverHeraguard ECO 0.9, 1.2, 1.5, 1.8, 1.8/95
51901077Work Surface WrapsStainless Steel Work SurfaceHeraguard ECO 1.8
Catalog number 50052462
Price (MYR)
-
Product Type:
Wind Screen Protection Kit
Description:
Windscreen protection kit
For Use With (Equipment):
Heraguard ECO 1.8
Integrate Thermo Scientific™ Heraguard™ ECO Clean Bench Accessories with the ECO clean bench to increase the flexibility and enhance operations. Options include media valves, solenoid valve, stainless-steel work surface wraps, side window modifications, UV and UV night covers, windscreen protection kits and additional accessories.
†Due to DC motors, Heraguard ECO clean benches use less energy to operate so there is less heat emitted into the lab than traditional AC motor clean benches. Based on data from internal testing and The University of Michigan Field Study, published in American Biotechnology Laboratory.
Specifications
For Use With (Equipment)Heraguard ECO 1.8
DescriptionWindscreen protection kit
Product TypeWind Screen Protection Kit
Configuration OptionsFactory Installed
Unit SizeEA