Flame Filter for Solvents
Flame Filter for Solvents
Actual product may vary
Flame Filter for Solvents
To be connected to vacuum port at rear wall, is recommended for applications with solvents with ignition temperatures of < 21 °C
Have Questions?
Catalog NumberFor Use With (Equipment)
50042626Vacutherm VT 6000 M/P-BL Oven, Vacuum Tubing
Catalog number 50042626
Price (MYR)
33,372.00
EA
Add to cart
For Use With (Equipment):
Vacutherm VT 6000 M/P-BL Oven, Vacuum Tubing
Price (MYR)
33,372.00
EA
Add to cart
To be connected to vacuum port at rear wall, is recommended for applications with solvents with ignition temperatures of < 21 °C
  • To be connected to vacuum port at rear wall, is recommended for applications with solvents with ignition temperatures of < 21°C.
Specifications
Product TypeFlame Filter
For Use With (Equipment)Vacutherm VT 6000 M/P-BL Oven, Vacuum Tubing
Unit SizeEA