Diaphragm Pump HMD 4C
Diaphragm Pump HMD 4C
Actual product may vary
Diaphragm Pump HMD 4C
Three Step Chemical Pump for Drying (Caustic/Corrosive Vapours). Supplied Complete with All Necessary Tubing and Fittings to Connect to the Vacuum Oven.
Have Questions?
Catalog number 50028362
Price (MYR)
86,889.00
EA
Add to cart
Price (MYR)
86,889.00
EA
Add to cart
Three Step Chemical Pump for Drying (Caustic/Corrosive Vapours). Supplied Complete with All Necessary Tubing and Fittings to Connect to the Vacuum Oven.
  • Ready to connect
  • Vacuum pump for vacuum drying ovens: quality, power and a long service life
  • Fully lined with PTFE and other fluorinated hydrocarbons
  • Functional design: integrated glass condensate trap, which empties quickly, and an emission condenser with cooling, supplied as standard
Specifications
Max. Vacuum3.20 kW
Certifications/ComplianceCE certified
Dimensions (L x W x H)338 x 241 x 500 mm
ModelHMD 4 C
Weight (Metric)18 kg
Flow Rate3.0 m3/h
Pump TypeVacuum Pump
Enclosure RatingIP20
ApplicationsFor Drying (Caustic/Corrosive Vapours)
Maximum Pressure<10 Mbar final pressure with gas ballast
For Use With (Equipment)Vacutherm 6000 Ovens in Particular The BL Model