E1-ClipTip™ Multichannel Pipettes
E1-ClipTip™ Multichannel Pipettes
Actual product may vary
E1-ClipTip™ Multichannel Pipettes
E1-ClipTip™ Multichannel Pipettes
E1-ClipTip™ Multichannel Pipettes
E1-ClipTip™ Multichannel Pipettes
E1-ClipTip™ Multichannel Pipettes
With enhanced versatility and customization, the Thermo Scientific™ E1-ClipTip™ Electronic Multichannel Pipette automates daily 96-and 384-well microplate pipetting tasks with the sealed in security of Thermo Scientific™ ClipTip™ technology.
Have Questions?
Catalog NumberVolume (Metric)Number of ChannelsCompatible Tips
467108010 to 300 μL12ClipTip 300
Catalog number 4671080
Price (MYR)
-
Volume (Metric):
10 to 300 μL
Number of Channels:
12
Compatible Tips:
ClipTip 300
With enhanced versatility and customization, the Thermo Scientific™ E1-ClipTip™ Electronic Multichannel Pipette automates daily 96-and 384-well microplate pipetting tasks with the sealed in security of Thermo Scientific™ ClipTip™ technology. Enjoy pipetting again with optimum functionality for every application and user preference. Feel the difference.
Innovative ClipTip interlocking tip interface technology
  • Tip is locked and sealed in place until ejected— no dropped tips that waste valuable samples
  • Complete seal to secure the sample volume in each tip for enhanced accuracy and precision
  • Tips are attached and ejected with a light touch— eliminates the frustration of banging tips on the pipette
An ideal choice for sample transfers
Reduce repetition and save time between labware formats with adjustable tip spacing in Equalizer models.
Unified operation for multiple users and environments
  • Personalized user interface and password protection ensure consistent operation among multiple users.
  • Electronic operation to help you enjoy pipetting again
  • Designed for optimum comfort and ease of use, the electronic tip ejection and index finger pipetting action let your thumb relax during pipetting.
Two operating options ensure optimal function for every application
Choose a step based Matrix or Presets with settings for your most common applications
MyPipette™ - One click access for more efficient and unified pipetting
Customize your main menu view by organizing icons in your preferred order or creating shortcuts for your favorite programs and pipetting functions. You can also protect important settings from unintentional editing with a password.
Store up to 20 programs
Tired of continuous editing? The Programs function allows you to store 20 your most common Matrix or Presets protocols with a specific name.
A pipette for every application
Choose from single channels, 8-16 channel multichannels, or 6- 12-channel adjustable tip spacing Equalizer multichannel pipettes within the volume range of 0.5-1250μL
Track your pipette
Service and calibration trackers remind you when the pipette needs to be sent to serviced.
Liquid specific calibrations
Higher quality results with liquid specific calibrations.
Li-Ion battery
Long lasting performance and fast recharging.
Flexible re-charging
Use either charging stand or a plug, you decide.
2-year warranty
With convenient web registration.
Compatible with Thermo Scientific ClipTip pipette tips.
†E1-ClipTip 12.5 and all 384 E1-ClipTip pipettes utilize snap and lip tip fitting technology.
Specifications
Accuracy±2.0 to 5.0%
Compatible TipsClipTip 300
DescriptionE1-ClipTip 12-channel
ColorOrange
For Use WithClipTip 300
ModelE1-ClipTip
Number of Channels12
Product LineE1-ClipTip
ProgrammabilityYes
TypeElectronic Pipette
Volume (Metric)10 to 300 μL
Increments0.1μL
Precision≤0.6 to 2.0%
Unit SizePK