Specifications
Accuracy0.5% (50μL)
Product TypeBottletop Dispenser
Angle90°
AutoclavableNot Autoclavable
Certifications/ComplianceISO 9001
ColorGray
DensityUp to 3.8 g/cm3
DescriptionWithout recirculation valve
Channels1
MaterialPlatinum Iridium Valve Spring
Volume (Metric)1 to 10 mL
DisplayAnalog
For Use With (Application)Dispensing acids, bases and hydrogen peroxide in the volume range from 1 to 10mL
IncludesDispensette S bottletop dispenser (for GL 45 threaded bottles), DE-M marking, performance certificate, telescoping filling tube, mounting tool, and boddle adapters GL 28-S 28 (ETFE, GL 32 (ETFE), and S40 (PTFE)
Increments0.2 mL
Precision0.1% (10μL)
Temperature (English)59°F to 104°F
Temperature (Metric)15°C to 40°C
Vapor PressureUp to 600 mbar
Viscosity RatingUp to 500 mm 2/s
Warranty2 Year
Unit SizeEA