S52-ST Swinging-Bucket Rotor for MTX and MX Plus Micro-Ultracentrifuges
S52-ST Swinging-Bucket Rotor for MTX and MX Plus Micro-Ultracentrifuges
Actual product may vary
S52-ST Swinging-Bucket Rotor for MTX and MX Plus Micro-Ultracentrifuges
Reach 4 x 5mL capacity at speeds up to 52,000rpm. Recommended for small particle separations such as nanoparticles, viral particles and density gradients.
Have Questions?
Catalog NumberCapacity
459774 x 5 mL
Catalog number 45977
Price (MYR)
101,242.00
EA
Add to cart
Capacity:
4 x 5 mL
Price (MYR)
101,242.00
EA
Add to cart
Reach 4 x 5mL capacity at speeds up to 52,000rpm with the Thermo Scientific™ S52-ST Swinging-Bucket Rotor. Recommended for small particle separations such as nanoparticles, viral particles and density gradients.
Features:
  • Maximum capacity: 4 × 5mL (0.02L)
  • Max. speed/RCF: 52,000rpm/275,458 x g
  • K factor: 78.6
  • Angle: 180°
Compatible with:
Thermo Scientific Sorvall MTX 150 Series Micro-Ultracentrifuge, MX Series Floor Model Micro-Ultracentrifuge
Recommended for:
Small particle separations such as nanoparticles and viral particles, density gradients.
Specifications
DescriptionS52-ST Swinging-Bucket Rotor
MaterialTitanium
TypeSwinging Bucket Rotor
Capacity4 x 5 mL
Max. RCF275,458 x g
Max. Speed52,000 rpm
Radius (Metric)91.2 mm
Radius (Metric) Min.39.4 mm
K Factor78.6
Max. Tube Length51 mm
Max. Tube Diameter13 mm
Net Weight3.7 lb. (1.7 kg)
For Use With (Equipment)Thermo Scientific™ Sorvall™ MTX and MX Plus micro-ultraCentrifuges
Angle180°
Avg. RCF197,230 x g
Min. RCF119,009 x g
Unit SizeEA