Search Thermo Fisher Scientific
Catalog Number | Capacity |
---|---|
45621 also known as 45/621 | 8 x 6.5 mL |
Ordering Information:
Order Ultracrimp/Clearcrimp Sealing Tool (03920), Crimp Gauge Replacement (03919), Extra Plugs and Caps (03999), Tube Cap Vise (52176), Tube Removal Tool (52177) separately. Contact your Sales Representative for information.
Includes:
Rotor stand, vacuum grease, lubricant, extra overspeed decal (100,000rpm), rotor cover O-ring (two extra), locking nut O-ring (two extra), rotor cover tool, utraspeed rotor log book, instruction manual
Compatible with:
Thermo Scientific Sorvall WX Ultra Series Centrifuge
Recommended for:
Protein and small virus isolation