S150-AT Fixed Angle Rotor
S150-AT Fixed Angle Rotor
Actual product may vary
S150-AT Fixed Angle Rotor
S150-AT Fixed Angle Rotor
S150-AT Fixed Angle Rotor
Perform high-speed applications with the Thermo Scientific™ S150-AT Fixed Angle Rotor, which operates at 150,000rpm with 8 x 2.0mL capacity.
Have Questions?
Catalog NumberCapacity
455828 x 2 mL
Catalog number 45582
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity:
8 x 2 mL
Perform high-speed applications with the Thermo Scientific™ S150-AT Fixed Angle Rotor, which operates at 150,000rpm with 8 x 2.0mL capacity.
Features:
  • Fixed angle (30°) titanium rotor
  • Capacity: 8 × 2.0mL
  • Maximum Speed/RCF: 150,000rpm/899,744 x g
  • K factor: 6.1
Compatible with:
Thermo Scientific™ Sorvall™ MTX 150 Series Micro-Ultracentrifuge, MX Series Floor Model Micro-Ultracentrifuge
Specifications
DescriptionS150-AT Fixed Angle Rotor
MaterialTitanium
TypeFixed Angle Rotor
Capacity8 x 2 mL
Max. RCF899,744 x g
Max. Speed150,000 rpm
Radius (Metric)35.8 mm
Radius (Metric) Min.20.8 mm
K Factor6.1
Max. Tube Length34 mm
Max. Tube Diameter11 mm
Net Weight1.1 lb. (0.49 kg)
For Use With (Equipment)Thermo Scientific™ Sorvall™ MTX and MX Plus micro-ultraCentrifuges
Angle30°
Avg. RCF711,250 x g
Min. RCF522,756 x g
Unit SizeEA