Search Thermo Fisher Scientific
Use the gold-standard Thermo Scientific™ LinerGOLD™ GC Liners for consistently high accuracy, sensitivity, and precision in your gas chromatography (GC) analysis.
Catalog Number | Diameter (Metric) Inner | Length (Metric) | Quantity | Product Type |
---|---|---|---|---|
453T1905 | 5 mm | 105 mm | 5 | Split FocusLiner |
45322044 | 2 mm | 120 mm | 2 | PTV Siltek Metal Liner |
45350030 | 5 mm | 105 mm | 5 | Split Straight Liner |
45352057 | 2 mm | 120 mm | 5 | PTV Straight Liner |
45352062 | 1 mm | 120 mm | 5 | PTV Liner, Three Baffles |
453T2120 | 2 mm | 120 mm | 5 | PTV Baffle Liner (Siltek) |
HYP453T2120 | 2 mm | 120 mm | 5 | PTV Baffle Liner (Siltek) |
45322045 | 2 mm | 120 mm | 5 | PTV Glass Straight Liner (Deactivated) |
453A1335-UI | 1 mm | 78.5 mm | 5 | Direct Straight Liner |
HYP453A1335-UI | 1 mm | 78.5 mm | 5 | Direct Straight Liner |
Use the gold standard in GC liner performance and consistency. LinerGOLD GC Liners provide enhanced inertness, leading to increased accuracy, sensitivity, and precision in your gas chromatography (GC) analysis. These injection port liners deliver:
LinerGOLD GC liners are manufactured and tested to ensure that you have confidence with every liner that is installed onto your GC system.