Injection Port Liners for GC Instruments
Injection Port Liners for GC Instruments
Actual product may vary
Injection Port Liners for GC Instruments
Use these injection port liners to optimize performance of Agilent GC instruments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric) InnerPacking MaterialProduct TypeQuantity
453A12954 mmNoneSplit Straight Liner5
HYP453A12954 mmNoneSplit Straight Liner5
453A13154 mmQuartz WoolSplit/Splitless FocusLiner w/ Single Taper5
453A13351.2 mmNoneDirect Straight Liner5
453A13454 mmNoneSplit/Splitless Liner w/ Single Taper5
453A19254 mmQuartz WoolSplitless Liner w/ Single Taper5
453A22654 mmQuartz WoolSplit Straight Liner5
HYP453A22654 mmQuartz WoolSplit Straight Liner5
Catalog number 453A1295
Price (MYR)
841.00
PK
Add to cart
Diameter (Metric) Inner:
4 mm
Packing Material:
None
Product Type:
Split Straight Liner
Quantity:
5
Price (MYR)
841.00
PK
Add to cart
Optimize the performance of Thermo Scientific™ and Agilent™ GC systems with our specifically designed Thermo Scientific™ Injection Port Liners for Agilent instruments. Highly deactivated and produced to exacting tolerances, these injection liners ensure exceptional reproducibility.
  • Highly deactivated borosilicate glass
  • Quartz wool packing material
  • IDxODxL: 4 x 6.3 x 78.5 mm
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description453A1295
Quantity5
Length (Metric)78.5 mm
Packing MaterialNone
Product TypeSplit Straight Liner
Diameter (Metric) Inner4 mm
Diameter (Metric) Outer6.3 mm
For Use With (Equipment)GC Injection Port
Liner MaterialHighly Deactivated Borosilicate Glass
Unit SizePK