Nunc™ 384-Well Polypropylene Sample Processing & Storage Microplates
Nunc™ 384-Well Polypropylene Sample Processing & Storage Microplates
Actual product may vary
Nunc™ 384-Well Polypropylene Sample Processing & Storage Microplates
Save on costs when storing compounds for small volume applications. Plates feature lower binding capacity so proteins or DNA will not bind, allowing for complete sample recovery.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorSterilityWell ShapeFit for Purpose
4312NaturalNon-sterileSquare-RoundedCertified Quality
4309NaturalSterileSquare, Conical Bottom (V-well)Certified Quality
Catalog number 4312
Price (MYR)
4,567.00
CS
Add to cart
Color:
Natural
Sterility:
Non-sterile
Well Shape:
Square-Rounded
Fit for Purpose:
Certified Quality
Price (MYR)
4,567.00
CS
Add to cart
Minimize costs when storing compounds for small volume applications. Thermo Scientific™ 384-Well Conical Bottom Polypropylene Plates feature a lower binding capacity so proteins or DNA will not bind, allowing for complete sample recovery. Withstands temperatures from -80° to +121°C. Offered in a variety of colors for quick identification of your storage plates.
  • Conical wells optimize sample recovery
  • Square rounded wells minimize wicking (capillary action)
  • Chemical resistant to DMSO
Specifications
BarcodeNo Barcode
Certifications/ComplianceCertified RNase and DNase Free. ANSI Compliant.
FormatStandard
Description384-well Polypropylene Sample Processing & Storage Plates
Array16 x 24
AutoclavableAutoclavable
Packaging10 pk., 80 cs.
Binding TypeLow binding PP
Product LineNunc
ColorNatural
Product Type384-well plate
For Use With (Equipment)ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers
MaterialPolypropylene
No. of Wells384
Volume (Metric) Working120 μL
SterilityNon-sterile
Surface TreatmentNon-treated
Well ShapeSquare-Rounded
Dnase FreeYes
For Use With (Application)Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation
No. per Case80
No. per Pack10
Regulatory StatusFor Research Use Only. Not for use in diagnostic procedures.
RNase FreeYes
Fit for PurposeCertified Quality
Plate SizeANSI/ SLAS Standard
Unit SizeCS