Alternate Cord and Plug Options for Ultra-Low Temperature Freezers
Actual product may vary
Alternate Cord and Plug Options for Ultra-Low Temperature Freezers
Meet your local requirements with Thermo Scientific™ Alternate Cord and Plug Options for Ultra-Low Temperature Freezers.
Have Questions?
Catalog Number
Type
Plug Type
Amperage
Voltage
430267
Twist Lock (Factory Installed)
US NEMA L6-15 P
15 A
230 V
Catalog number 430267
Price (MYR)
1,244.00
EA
Add to cart
Type:
Twist Lock (Factory Installed)
Plug Type:
US NEMA L6-15 P
Amperage:
15 A
Voltage:
230 V
Price (MYR)
1,244.00
EA
Add to cart
Meet your local requirements with Thermo Scientific™ Alternate Cord and Plug Options for Ultra-Low Temperature Freezers. Options include an array of standard and special plugs for North America and international markets.
Specifications
TypeTwist Lock (Factory Installed)
For Use With (Equipment)Thermo Scientific™ TDE Series, Forma™ 900/700, HERAfreeze HFU B, Revco™ ExF/DxF, Thermo Scientific™ TSE Series