Nalgene™ Large HDPE Funnels
Nalgene™ Large HDPE Funnels
Actual product may vary
Nalgene™ Large HDPE Funnels
Nalgene™ Large HDPE Funnels
Nalgene™ Large HDPE Funnels
Nalgene™ Large HDPE Funnels
Choose a safer alternative to glass with this shatterproof funnel, featuring external and internal ribbing for air release.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)TopCapacity (Metric)
4262-0110258mm4.06 L
4262-0080201mm1.9 L
Catalog number 4262-0110
Price (MYR)
1,988.00
CS
Add to cart
Diameter (Metric)Top:
258mm
Capacity (Metric):
4.06 L
Price (MYR)
1,988.00
CS
Add to cart
Replace glass and metal lab funnels with Thermo Scientific™ Nalgene™ Large HDPE Funnel, featuring external and internal ribbing for air release. These funnels are chemically resistant to a wide variety of lab chemicals and reagents and are both shatterproof and dentproof making them a valuble asset to lab safety programs.
  • High-density polyethylene is shatterproof and dentproof, making it a reliable long-term tool for the lab
Recycling Information:
Nalgene reusable labware products are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Thermo Fisher Scientific encourages recycling High Density Polyethylene (HDPE) labware at end of product life. Please also recycle the corrugate cartons and any plastic bags (LDPE) included in your product packaging.
Specifications
Diameter (Metric)Top258mm
Capacity (Metric)4.06 L
Diameter (Metric) Stem25 mm
MaterialHDPE
Height (Metric)301 mm
AutoclavableNot Autoclavable
DescriptionFunnel Large HDPE 279mm
Length (Metric) Stem90 mm
Quantity1/Pk.; 6/Cs.
Unit SizeCS