RadEye™ B20 and B20-ER Multi-Purpose Survey Meters
Actual product may vary
RadEye™ B20 and B20-ER Multi-Purpose Survey Meters
Quickly characterize mixed surface contamination from gamma radiation, x-ray radiation, alpha radiation and beta radiation with dose rate measurement survey meters.
Have Questions?
Catalog NumberDescription
425068510RadEye B20-ER Multi-Purpose Survey Meter
Catalog number 425068510
HSN 90301000
Price (MYR)
-
Description:
RadEye B20-ER Multi-Purpose Survey Meter
First responders need to quickly identify mixed radioactive surface contamination in facility and field environments. Thermo Scientific™ RadEye™ B20 and B20-ER Multi-Purpose Survey Meters are simple, robust, reliable contamination and dose rate measurement tools for characterizing alpha, beta, gamma and X-ray radiation. RadEye B20 and B20-ER Multi-Purpose handheld Geiger counters provide an on-site solution for homeland security personnel, fire brigades, emergency response personnel, agencies involved in decontamination and decommissioning projects and hospital and pharmaceutical industry employees.
RadEye B20 and B20-ER Multi-Purpose Survey Meters offer consistent, reliable results in any application requiring radiological surveys of flat surfaces. Model B20 is for normal measurements. Model B20-ER is for high-range measurements.
  • Lightweight (300g), excellent grip with and without gloves.
  • Rugged and compact design, thick rubber protective cover.
  • Low cost of ownership with > 500 hour operation time with 2 AAA batteries – rechargeable NiMH-cells can be used.
  • Menu-driven user interface results in low training cost and immediate familiarity.
  • Huge internal data memory for both scaler results and continuous data recording.
  • Bright backlit LCD display. Plain text messages and different languages can be selected.
  • Easy adaptation to different tasks by supervisor configuration, calibration, selection of measuring units.
  • Versatile operation modes:
    • Scaler/Timer with preset count and preset time for sample measurements.
    • Continuous ratemeter mode for frisker operation.
    • Dose rate mode.
  • Audible indication: single pulse or chirper mode proportional to count rate.
  • Earphone output for operation in loud environment.
  • IR PC Interface or Bluetooth™ as option.
  • Advanced Windows™ software is available as option.
View our online training here.
Specifications
Sensitivity0 to 100 mSv/h
DescriptionRadEye B20-ER Multi-Purpose Survey Meter
Dimensions (L x W x H)5.2 x 2.8 x 5.2 in. (13 x 7 x 13 cm)
Diameter (English) Window1.7 in.
Energy Range17 keV to 3 MeV; according to IEC 60846-1)
Battery Type2 AAA
Product TypeSurvey Meter
Measurement RangeContamination B-20: 0 - 10 kcps; B20ER: 0 - 500 kcps
Height (English)5.2 in.
Length (English)5.2 in.
Efficiency2p Efficiency (ref. to 50 mm diameter without rubber sleeve): Am-241; Co-60: 25%; Sr/Y-90: 36%; C-14: 19%
Detector TypePancake GM-tube, window dia. 44 mm (1.7 in.) 1.8 to 2.0mg/cm2
Diameter (Metric) Window44 mm
Width (Metric)7 cm
Weight (Metric)0.3 kg
For Use With (Application)Civil defense, fire brigades, hospitals, nuclear industry, and pharmaceutical industry
Alarm TypesLED, sound, vibrator
Radiation TypeGamma
Unitscps, cpm. Bq, dpm, dps, Sv/h, rem/h
Depth (Metric)6 mm
Width (English)2.8 in.
Battery Life>500 hr.s on two AAA batteries
Dose Rate Limit100 mSv/h (10 rem/h)
Weight (English)0.7 lb.
Length (Metric)13 cm
Depth (English)2.4 in.
Height (Metric)13 cm
Dose RangeGamma Dose Rate: 0 to 100 mSv/h (0 to 10 rem/h); Contamination: 0 to 500 kcps
Unit SizeEach