Nalgene™ PMP Erlenmeyer Flasks with Closure
Nalgene™ PMP Erlenmeyer Flasks with Closure
Actual product may vary
Nalgene™ PMP Erlenmeyer Flasks with Closure
Nalgene™ PMP Erlenmeyer Flasks with Closure
Nalgene™ PMP Erlenmeyer Flasks with Closure
Nalgene™ PMP Erlenmeyer Flasks with Closure
Perform titrations, preparation and storage of culture media with these flasks, which provide a combination of chemical resistance, strength and transparency.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
4109-10001000 mL53 mm
4109-0250250 mL38 mm
4109-0500500 mL43 mm
Catalog number 4109-1000
Price (MYR)
1,236.00
CS
Add to cart
Capacity (Metric):
1000 mL
Closure Size:
53 mm
Price (MYR)
1,236.00
CS
Add to cart
Perform titrations, preparation and storage of culture media with Thermo Scientific™ Nalgene™ PMP Erlenmeyer Flasks with Closure, which provide a combination of chemical resistance, strength and transparency. Rugged, shatter-proof construction combined with light weight make these flasks a valuble element of lab safety programs.
Features:
  • Excellent chemical resistance to a wide variety of lab reagents
  • More transparent than polypropylene but with similar chemical resistance
  • Ideal for preparation and storage of culture media and titrations
  • All sizes have easy-to-read graduations
  • Autoclavable/Graduated
Includes:Polypropylene screw closures
.
Specifications
Capacity (English)32 oz.
DescriptionFlask Erlenmeyer Screw Closure PMP 1000mL
Capacity (Metric)1000 mL
MaterialPMP
No. per Case6
No. per Pack2
AutoclavableAutoclavable
DisposableReusable
GraduatedYes
Mouth StyleNarrow
SterilityNon-sterile
Bottom ShapePlain
Closure TypeScrew
Closure Size53 mm
Unit SizeCS