Nunc™ Deep Form Bacteriology Petri Dishes
Nunc™ Deep Form Bacteriology Petri Dishes
Actual product may vary
Nunc™ Deep Form Bacteriology Petri Dishes
Culture fungi, bacteria and other microorganisms with these petri dishes
Have Questions?
Catalog NumberDiameter (Metric)
4014-12150 mm
Catalog number 4014-12
Price (MYR)
1,423.00
CS
Add to cart
Diameter (Metric):
150 mm
Price (MYR)
1,423.00
CS
Add to cart
Culture fungi, bacteria and other microorganisms with Thermo Scientific™ Nunc Lab-Tek Disposable Petri Dishes.
Thermo Scientific Nunc Cell Culture products offer solutions for cell growth and analysis and have been used by researchers worldwide since 1953. With a wide range of surface areas and treatments, we have products for all your basic or advanced cell-based research.
Features:
  • Promote larger plaques used for efficient library screening
  • Compatible with automated systems
  • Sterile; disposable
  • Deep Petri dishes allow for longer culture periods
Ordering Information:
12 per sealed plastic sleeve.
Specifications
DisposableSingle-use
ShapeRound
SterilitySterile
TypePetri Dish
VentNo
Height (Metric)26 mm
Diameter (Metric)150 mm
Unit SizeCS