Continuous Operation Rocking and Rolling Action Roller Shakers
Continuous Operation Rocking and Rolling Action Roller Shakers
Actual product may vary
Continuous Operation Rocking and Rolling Action Roller Shakers
Roller shakers with six or ten rollers for a wider range of applications, especially in the medical and biological field. IKA™ Continuous Operation Rocking and Rolling Action Roller Shakers provide smooth rocking and rolling action at infinitely adjustable speeds.
Have Questions?
Catalog NumberModel
4012-000Roller 10 Basic
Catalog number 4012-000
Price (MYR)
1,715.00
EA
Add to cart
Model:
Roller 10 Basic
Price (MYR)
1,715.00
EA
Add to cart
  • Used for mixing blood samples, solid and liquid suspensions and viscous samples 
  • Unit can be used with all conventional tubes and cylindrical bottles
  • Easily removable attachments for quick cleaning in case of sample spills
  • Timer: Countdown, adjustable from 1 sec to 99 hr. and 59 min.
  • Counter: Display of shaking time
  • Error Code Display
  • Usable at 4 to 50°C in incubators
  • Suitable for continuous operation
  • Removable rolls enable shaking of larger tubes
  • Easy operation with keypad
  • Shaking diameter: 24.5mm (height)
  • Speed range: 5 to 80rpm
Specifications
Rotating Angle
Speed Range30 rpm
Voltage100/240 V
Temperature Range (Metric)4°C to 50°C
Wattage24 W
Certifications/ComplianceDIN EN 60529 IP 21
Frequency50/60 Hz
Height (Metric)115 mm
Length (Metric) Platform350 mm
Shipping Weight (Metric)6 kg
ModelRoller 10 Basic
For Use With (Application)Used for mixing blood samples, solid and liquid suspensions and viscous samples
Humidity Range80%
Packaging Quantity1 Ea.
Unit SizeEA