Check Order Status
Status H. Pylori Test Kit
Actual product may vary
Status H. Pylori Test Kit
Rapid immunoassay for the qualitative detection of the anti-helicobacter pylori antibody in human whole blood, serum or plasma
Have Questions?
Catalog Number
Quantity
37030
30 Tests
Catalog number
37030
Price (MYR)
-
Quantity:
30 Tests
Features:
CLIA waived for whole blood
Sensitivity of 95.9% with 91.6% overall agreement to biopsy histology
Easy-to-use single reagent procedure
Results available in 10 minutes
Requires only 25μL of whole blood or 10μL serum/plasma
Convenient fingerstick sample collection
Specifications
Accuracy
91.6% overall agreement to biopsy histology
Detectable Analytes
Anti-helicobacter pylori antibodies
Sample Type
Whole Blood, Serum, Plasma
Type
H. pylori Immunoassay
Quantity
30 Tests
Test Time
10 min.
Unit Size
Foot