Thermo Scientific™ Pre-cut PEEK Tubing in a 5-foot Coil is easily cut to the desired length.
Have Questions?
Change view
Catalog Number
Diameter (English) Inner
37007
0.007 in.
37040
0.040 in.
2 Options
Catalog number 37007
Price (MYR)
-
Diameter (English) Inner:
0.007 in.
Thermo Scientific™ Pre-cut PEEK Tubing in a 5-foot Coil is easily cut to the desired length.
Broad chemical compatibility
Biocompatible
May be used with standard fingertight fittings
Resistant to most organic solvents however nitric acid, sulfuric acid, dichloromethane, THF and DMSO are not recommended
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.