Nalgene™ Double-Scale PMP Pharmaceutical Graduates
Nalgene™ Double-Scale PMP Pharmaceutical Graduates
Actual product may vary
Nalgene™ Double-Scale PMP Pharmaceutical Graduates
Nalgene™ Double-Scale PMP Pharmaceutical Graduates
Nalgene™ Double-Scale PMP Pharmaceutical Graduates
Nalgene™ Double-Scale PMP Pharmaceutical Graduates
Avoid breakage with transparent and shatterproof double-scale PMP pharmaceutical graduates. Lighter in weight than glass, they provide excellent chemical resistance.
Have Questions?
Catalog NumberCapacity (Metric)Limit of Error
3673-0004125 mL±1.8 mL
Catalog number 3673-0004
Price (MYR)
1,588.00
CS
Add to cart
Capacity (Metric):
125 mL
Limit of Error:
±1.8 mL
Price (MYR)
1,588.00
CS
Add to cart
Avoid breakage with transparent and shatterproof Thermo Scientific™ Nalgene™ Double-Scale PMP Pharmaceutical Graduates. Lighter in weight than glass, they provide excellent chemical resistance. They are graduated in ounces and milliliters with no meniscus to confuse readings.
  • Meet the requirement of ASTM E1094
  • Autoclavable (will affect accuracy)
  • Graduated
Specifications
Base TypeHexagonal
ScaleDual Scale mL and ounces
Capacity (Metric)125 mL
Certifications/ComplianceASTM E1094
Height (English)5.84 in.
Height (Metric)148.3 mm
MaterialPMP
Limit of Error±1.8 mL
Capacity (English)4 oz.
No. per Pack1
No. per Case12
Capacity4 oz., 125 mL
AutoclavableAutoclavable
Diameter (English)Top2.39 in.
Diameter (Metric)Top60.7 mm
Product TypeGraduated Cylinder
ShapeConical
Tolerance±1.8 mL
TypePharmaceutical Graduate
DescriptionGraduated Cylinder Pharmaceutical PMP 125mL
Unit SizeCS