Male Luer-LOK Priming Syringes
Male Luer-LOK Priming Syringes
Actual product may vary
Male Luer-LOK Priming Syringes
Thermo Scientific™ Male Luer-LOK Priming Syringes offer easy, accurate and reproducible manual injections.
Have Questions?
Catalog NumberVolume (Metric)Product Type
365N272110 mLGC Autosampler Syringe
Catalog number 365N2721
Price (MYR)
-
Volume (Metric):
10 mL
Product Type:
GC Autosampler Syringe
Thermo Scientific™ Male Luer-LOK Priming Syringes offer easy, accurate and reproducible manual injections.
  • Square tip to prevent damage to the injector
  • Wide range of volumes
  • Precision made from borosilicate glass and stainless steel
  • Robust design and easy-to-read markings
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description10 mL FN GT LC, 19 G, 57 mm
Needle Gauge19 G
Length (Metric)57 mm
Product TypeGC Autosampler Syringe
QuantityEa.
Volume (Metric)10 mL
Length (Metric) Needle57 mm
Needle Point StyleLC
For Use With (Equipment)TriPlus RSH Autosampler
PTFE Tipped PlungerYes
Unit SizeEach