Manual, Removable-Needle, Gas Tight Syringes for GC Instruments
Manual, Removable-Needle, Gas Tight Syringes for GC Instruments
Actual product may vary
Manual, Removable-Needle, Gas Tight Syringes for GC Instruments
Use Thermo Scientific™ Removable Needle Syringes for accurate and reproducible manual sample introduction into GC instruments, with or without the needle.
Have Questions?
Catalog NumberVolume (Metric)Needle GaugeNeedle Point Style
365DLG2110 μL22 G90° Blunt
Catalog number 365DLG21
Price (MYR)
-
Volume (Metric):
10 μL
Needle Gauge:
22 G
Needle Point Style:
90° Blunt
Get durability, clarity and accuracy from Thermo Scientific™ Manual Removable Needle Syringes. Used for manual sample injection into gas chromatography (GC) instruments with or without the needle, they provide accuracy and reproducibility that provides confidence in analytical results, time-after-time. Choose from a variety of sizes, and types. These syringes are available in standard industry size with black easy-to-read and solvent-resistant markings.
  • Square tip to prevent damage to the injector
  • Wide range of volumes
  • Precision made from borosilicate glass and stainless steel
  • Robust design and easy-to-read markings
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Needle Gauge22 G
Length (Metric)50 mm
Product TypeGas Tight LC Autosampler Syringe, Removable Needle
DisposableReusable
Needle Point Number365 RNL15
Quantity1/Ea.
Volume (Metric)10 μL
Length (Metric) Needle50 mm
Needle Point Style90° Blunt
For Use With (Equipment)Rheodyne and Valco Injectors
Replacement Needle365RNL15
Unit SizeEach