GC Syringes for Agilent Instruments
GC Syringes for Agilent Instruments
Actual product may vary
GC Syringes for Agilent Instruments
Easily accomplish accurate, precise, and reproducible injections with Thermo Scientific™ GC Syringes for Agilent Instruments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Description
365D163610 μLTapered
365D162110 μLTapered
Catalog number 365D1636
Price (MYR)
-
Volume (Metric):
10 μL
Description:
Tapered
Achieve accurate, precise and reproducible injections easily using Thermo Scientific™ GC Syringes designed specifically for Agilent™ GC systems. Optimized for septum piercing, the rough, field-tested syringes are made of borosilicate glass with metal flanges of either stainless steel or nickel-plated brass.
  • Accurate, precise and reproducible over a range of volumes
  • Fixed needle syringes have a maximum temperature range of 5° to 70°C
  • 5 to 100μL capacity needles have a maximum safe working pressure of 1000psi; 100psi maximum functional pressure
  • Cone point needle tip to prevent plunger binding with dirty samples
  • 42mm length
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionTapered
Needle Gauge23 to 26s G
Length (Metric)42 mm
Product TypeGC Autosampler Syringe, Fixed Needle
DisposableReusable
Quantity6 Pk.
Volume (Metric)10 μL
Length (Metric) Needle42 mm
Needle Point StyleCone
For Use With (Equipment)Agilent GC Instruments
Unit SizeEach