ART™ Non-Filtered, Wide-Bore, Genomic Pipette Tips
ART™ Non-Filtered, Wide-Bore, Genomic Pipette Tips
Actual product may vary
ART™ Non-Filtered, Wide-Bore, Genomic Pipette Tips
Thermo Scientific™ ART™ Non-filtered, wide bore pipette tips are ideal for pipetting in genomic workflows.
Have Questions?
Catalog NumberVolume (Metric)SterilityFormat
35921000 μLNon-sterileLift-off Lid Rack
Catalog number 3592
Price (MYR)
1,490.00
CS
Add to cart
Volume (Metric):
1000 μL
Sterility:
Non-sterile
Format:
Lift-off Lid Rack
Price (MYR)
1,490.00
CS
Add to cart
Thermo Scientific™ ART™ Non-filtered, wide bore pipette tips are ideal for pipetting in genomic workflows. The wide orifice prevents shearing of cells and DNA and RNA creating longer, continuous strands. The wide opening also allows for easier pipetting of viscous fluids or solutions. Ideal for use in reference labs and blood banking applications.
  • Wide orifice is larger than standard tip openings making it ideal for pipetting fragile cell lines and viscous samples
  • Ideal for genomic work
  • Universal Tip, fits most brands of pipettes
  • ART™ non-filtered tips deliver the same proven quality, performance and reliability as ART™ barrier
  • Non-filtered tip
  • ART™ tips are DNase, RNase, DNA, ATP, PCR, and Endotoxin Free
  • Lift-off lid racks provide access to all tips, great for multichannel pipetting
Specifications
DescriptionART 1000G, Non-filtered, non-sterile, Racked, Lift-off Lid
Volume (Metric)1000 μL
FilteredNon-filtered
SterilityNon-sterile
ColorClear
AutoclavableAutoclavable
ModelART 1000G
Clean ClaimsDNase, RNase, DNA, ATP, Bioburden, PCR Inhibitor, Endotoxin, and Pyrogen Free
For Use With (Application)Genomic workflows, fragile cell lines, viscous liquids
FeaturesWide bore orifice
FormatLift-off Lid Rack
Tip StyleWide Orifice
Packaging3200/Pk.
Quantity3200/Pk.
DisposableSingle-use
For Use With (Equipment)Universal Fit
Product LineART
MaterialVirgin Polypropylene
Unit SizeCS