Nuts for Capillary GC Columns Connections
Nuts for Capillary GC Columns Connections
Actual product may vary
Nuts for Capillary GC Columns Connections
Use Thermo Scientific™ nuts with compatible ferrules to eliminate leaks and connect your capillary GC columns to your instrument.
Have Questions?
Catalog NumberDescriptionFor Use With (Equipment)
35050458
also known as HYP35050458
Retaining Nut, Hexagonal, 1/4-in. (M6) for GC capillary columnsTrace 1300 Series GC (SSL, FID, NPD, TCD, ECD, FPD) and Agilent GC
Catalog number 35050458
also known as HYP35050458
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
Retaining Nut, Hexagonal, 1/4-in. (M6) for GC capillary columns
For Use With (Equipment):
Trace 1300 Series GC (SSL, FID, NPD, TCD, ECD, FPD) and Agilent GC
Use Thermo Scientific™ nuts with compatible ferrules to eliminate leaks and connect your capillary GC columns to your instrument. Thermo Scientific™ ferrules are available in three different materials - 100% Graphite, 15% Graphite / 85% Vespel, or SilTite Metal - and various dimensions to match your instrument and capillary column inner diameter. Use the nut that is compatible with your ferrule.
Specifications
DescriptionRetaining Nut, Hexagonal, 1/4-in. (M6) for GC capillary columns
For Use With (Equipment)Trace 1300 Series GC (SSL, FID, NPD, TCD, ECD, FPD) and Agilent GC
MaterialStainless Steel
TypeCapillary Column Nut
QuantityPack of 5
Unit SizePack