Check Order Status
Spatula with Stainless-Steel Blade
Actual product may vary
Spatula with Stainless-Steel Blade
Collect, transfer, or stir chemical samples with ease. Usbeck™ Spatula with Stainless-Steel Blade is perfect for collecting, transferring, and stirring chemical samples.
Have Questions?
Catalog Number
Length (Metric)
Length (Metric) Blade
3500
155 mm
75 mm
Catalog number
3500
Price (MYR)
2,085.00
CS
Add to cart
Length (Metric):
155 mm
Length (Metric) Blade:
75 mm
Price (MYR)
2,085.00
CS
Add to cart
Stainless steel
Highly flexible
Polished, wooden handle
Specifications
Product Type
Spatula
Length (Metric) Blade
75 mm
Material
Stainless Steel
Length (Metric)
155 mm