Nunc™ Disposable Plastic Centrifuge Tubes
Nunc™ Disposable Plastic Centrifuge Tubes
Actual product may vary
Nunc™ Disposable Plastic Centrifuge Tubes
Nunc™ Disposable Plastic Centrifuge Tubes
Nunc™ Disposable Plastic Centrifuge Tubes
Nunc™ Disposable Plastic Centrifuge Tubes
Maintain a high level of accuracy while increasing lab safety with these disposable plastic centrifuge tubes, which are an excellent alternative to glass.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Bottom ShapeSterility
34314112 mLRoundSterile
34291912 mLRoundNon-sterile
Catalog number 343141
Price (MYR)
1,144.00
CS
Add to cart
Capacity (Metric):
12 mL
Bottom Shape:
Round
Sterility:
Sterile
Price (MYR)
1,144.00
CS
Add to cart
Maintain a high level of accuracy while increasing lab safety with Thermo Scientific™ Nunc™ Disposable Plastic Centrifuge Tubes, which are an excellent alternative to glass. Selected tubes are relative centrifugal force (RCF) tested. Nunc Labware Products are made from high purity resins and molded using our state of the art processes.
Features:
  • Polyethylene (PE) and polypropylene (PP) tubes are particularly useful in serology where the low adsorption of proteins is of great importance
  • Polystyrene (PS) tubes are transparent and suitable for a broad range of applications
  • Polyethylene and polypropylene tubes have good chemical resistance and high mechanical strength
  • Different cap types are available for a variety of applications and fit most standard centrifuges
Includes:
Cat. nos. 343141, 345608 supplied with mounted stoppers for sterile work.
Specifications
Capacity (Metric)12 mL
DisposableSingle-use
MaterialPolystyrene
No. per Case3600
SterilitySterile
ShapeRound
Bottom ShapeRound
Graduations5 and 10 mL
Product TypeCentrifuge Tube
Max. RCF3000 x g
Unit SizeCS