Check Order Status
Closure for 23 ml Oakridge Tubes
Closure for 23 ml Oakridge Tubes
Aluminum sealing cap for 23ml oakridge tubes
Have Questions?
Catalog Number
Capacity (Metric)
314347
23 mL
Catalog number
314347
Price (MYR)
500.00
EA
Add to cart
Capacity (Metric):
23 mL
Price (MYR)
500.00
EA
Add to cart
Aluminum sealing cap for 23ml oakridge tubes
Specifications
Capacity (Metric)
23 mL
Material
Aluminum
For Use With
23 mL Oakridge Tubes
Product Type
Sealing Cap