Septa
Septa
Actual product may vary
Septa
Septa
Septa
Septa
Septa
Septa
A wide range to cover most applications
Have Questions?
Catalog NumberDiameter (Metric) Inner
3130323311 mm
Catalog number 31303233
Price (MYR)
-
Diameter (Metric) Inner:
11 mm
A wide range to cover most applications
  • Variety of materials and temperature limits for all applications
  • For use in Thermo Scientific TRACE™ and FOCUS™ GCs and instruments from other manufacturers
  • Contaminant-free in individual blister packs
Specifications
For Use With (Equipment)GC Instruments
MaterialSilicone
TypeLow Bleed Septum
Diameter (Metric) Inner11 mm
QuantityPack of 50
Unit SizePack