VisionMate™ SR Single Rack 2D Barcode Reader
VisionMate™ SR Single Rack 2D Barcode Reader
Actual product may vary
VisionMate™ SR Single Rack 2D Barcode Reader
Decode a full rack of barcoded storage tubes in less than ten seconds with the compact, user-friendly Thermo Scientific™ VisionMate™ SR Single Rack 2D Barcode Reader.
Have Questions?
Catalog NumberDescription
3115-11
also known as 3115
VisionMate SR 2-D Bar Code Reader
Catalog number 3115-11
also known as 3115
Price (MYR)
42,313.00
EA
Add to cart
Description:
VisionMate SR 2-D Bar Code Reader
Price (MYR)
42,313.00
EA
Add to cart
Decode a full rack of barcoded storage tubes in less than ten seconds with the Thermo Scientific™ VisionMate™ SR Single Rack 2D Barcode Reader. This compact, user-friendly reader provides exceptional flexibility with data export formats, tube type readability, and tube layout options.
  • Controlled via on-screen software interface or remotely via TCP/IP connection
  • Scans a full tube rack in less than ten seconds
  • Simply follow the instructions during software installation and connect the reader to a computer via USB port
  • Integrates easily with LIMS, existing databases or tracking systems
  • Small footprint and compact profile save valuable laboratory space
Specifications
DescriptionVisionMate SR 2-D Bar Code Reader
Decoding CapacitySingle Rack
Data Transfer MethodSoftware Interface or Remotely via TCP/IP Connection
SpeedFull tube rack in less than ten seconds
TypeBarcode Reader
Unit SizeEA