Hypersil™ Phenyl HPLC Columns
Hypersil™ Phenyl HPLC Columns
Actual product may vary
Hypersil™ Phenyl HPLC Columns
Hypersil™ Phenyl HPLC Columns
Hypersil™ Phenyl HPLC Columns
Use as a general-purpose column with aromatic selectivity.
Have Questions?
Catalog NumberLength (Metric)Diameter (Metric)Particle Size
30903-154630150 mm4.6 mm3 μm
Catalog number 30903-154630
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
150 mm
Diameter (Metric):
4.6 mm
Particle Size:
3 μm
Effectively separate aromatic compounds and moderately polar compounds with Thermo Scientific™ Hypersil™ Phenyl LC Columns.
Classical Hypersil stationary phases are recognized as an industry standard in HPLC. Proven as an effective analytical tool, classical Hypersil phases are well established and referenced in HPLC methods. Quality is assured by manufacture under ISO 9001-2000 quality guidelines.
For more challenging separations that require improved chromatographic performance, use Thermo Scientific™ Hypersil GOLD™ Phenyl columns as an alternative.
  • For the separation of certain aromatic compounds and moderately polar compounds
  • Retention characteristics similar to Hypersil MOS
  • Alternative selectivity to C8
Ordering Information:
Other column dimensions are available. Also available in 10μm. Please call Customer Service for more information.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
For Use WithHPLC
Length (Metric)150 mm
Max. Pressure5800 psi (400 bar)
Particle Size3 μm
pH2 to 8
Pore Size120 Å
Surface Area170 m²/g
Temperature60°C
USP TypeL11
Stationary PhasePhenyl (Ph)
Product LineHypersil Classical
Packing MaterialSpherical, Fully Porous Type A Silica
Carbon Load5%
Column TypeReversed Phase
EndcappedNo
PhaseReversed Phase
Unit SizeEach