Hypersil™ SAS C1 LC Column
Hypersil™ SAS C1 LC Column
Actual product may vary
Hypersil™ SAS C1 LC Column
Separate moderately polar and multi-functional compounds with Thermo Scientific™ Hypersil™ SAS C1 LC columns.
Have Questions?
Catalog NumberLength (Metric)
30505-204630200 mm
Catalog number 30505-204630
Price (MYR)
-
Length (Metric):
200 mm
  • A short alkyl chain reversed phase material
  • Selectivity for polar and multifunctional compounds
  • Can be used in ion-pair separations
Specifications
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
Length (Metric)200 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size120 Å
Surface Area170 m²/g
Temperature60°C
USP TypeL13
Stationary PhaseC1
Packing MaterialSpherical, Fully Porous Type A Silica
Carbon Load2.5%
Column TypeMixed Mode
EndcappedNo
PhaseReversed Phase and Ion-Pairing
Unit SizeEach