Hypersil™ MOS C8 HPLC Columns
Hypersil™ MOS C8 HPLC Columns
Actual product may vary
Hypersil™ MOS C8 HPLC Columns
Use as a general-purpose C8 phase for acidic and neutral analytes.
Have Questions?
Catalog NumberLength (Metric)Diameter (Metric)Particle Size
30205-254630250 mm4.6 mm5 μm
Catalog number 30205-254630
HSN 90279090
Price (MYR)
0.00
Each
Add to cart
Length (Metric):
250 mm
Diameter (Metric):
4.6 mm
Particle Size:
5 μm
Price (MYR)
0.00
Each
Add to cart
For reliable, affordable analysis of more hydrophobic analytes such as small peptides and proteins, pharmaceuticals, steroids, and environmental samples, use Thermo Scientific™ Hypersil™ MOS C8 HPLC Columns.
Recognized as an HPLC industry standard, Hypersil stationary phases are proven effective analytical tools. Classical Hypersil phases are well established and referenced in HPLC methods. Quality is assured by manufacture under ISO 9001-2000 guidelines.
For separations requiring improved chromatographic performance, use Thermo Scientific™ Hypersil GOLD™ C8 columns as an alternative.
  • Reliable, less retentive phase
  • Long column lifetimes, even under basic conditions
  • Manufactured under ISO 9001-2000 quality guidelines
  • Affordable
Ordering Information:
Other column dimensions are available. Also available in 10 μm. Please call Customer Service for more information.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
Length (Metric)250 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size120 Å
Surface Area170 m²/g
Temperature60°C
USP TypeL7
Stationary PhaseC8
Product LineHypersil Classical
Packing MaterialSpherical, Fully Porous Type A Silica
Carbon Load6.5%
Column TypeReversed Phase
PhaseReversed Phase
Unit SizeEach